1-Ethynylcyclohexanol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L12591-22
L12591-36
CAAAAL12591-22EA
59.88
CAD
CAAAAL12591-22
CAAAAL12591-36
1-Ethynylcyclohexanol 99%
1-Ethynylcyclohexanol
Purity: 99%
CAS No.: 78-27-3
Molecular Formula: C8H12O
Formula Weight: 124.18
Flash Point: 73°(163°F)
Refractive Index: 1.4830
Odor: Mild
Melting Point: 30-33°
Boiling Point: 180°
Beilstein Reference: 471404
UN No.: 2811
MDL No.: MFCD00003858
Formula:
C₈H₁₂O Boiling Pt: 180 °C Melting Pt: 30…33 °C Density: 0.973 Flash Pt: 73 °C (163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003858 CAS Number: 78-27-3 EINECS: 201-100-9 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...