4-Iodophenyl isothiocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L12583-04
L12583-09
CAAAAL12583-04EA
50.4
CAD
CAAAAL12583-04
CAAAAL12583-09
4-Iodophenyl isothiocyanate 97%
4-Iodophenyl isothiocyanate
Purity: 97%
CAS No.: 2059-76-9
Molecular Formula: C7H4INS
Formula Weight: 261.09
Odor: Pungent
Melting Point: 74-78°
Beilstein Reference: 2412143
UN No.: 2923
MDL No.: MFCD00041091
Formula:
C₇H₄INS Boiling Pt: 296 °C (1013 hPa) Melting Pt: 74…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041091 CAS Number: 2059-76-9 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...