3-Hydroxypropiononitrile 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Ethylene Cyanohydrin, 2-Cyanoethanol, 3-Hydroxypropionitrile, Hydracrylonitrile, β-Hydroxypropiononitrile
L12570-0B
L12570-14
L12570-30
CAAAAL12570-0BEA
208.56
CAD
CAAAAL12570-0B
CAAAAL12570-14
CAAAAL12570-30
3-Hydroxypropiononitrile 97%
3-Hydroxypropiononitrile
Purity: 98%
CAS No.: 109-78-4
Molecular Formula: C3H5NO
Formula Weight: 71.08
Flash Point: 128°(262°F)
Refractive Index: 1.4250
Melting Point: -46°
Boiling Point: 227-228°
Beilstein Reference: 635773
Merck Reference: 14,3794
MDL No.: MFCD00002826
Formula:
C₃H₅NO MW: 71.08 g/mol Boiling Pt: 227…228 °C Melting Pt: –46 °C Density: 1.041 Flash Pt: 128 °C(262 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002826 CAS Number: 109-78-4 EINECS: 203-704-8 Merck Index: 14,3794 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...