Sodium dihydrogen citrate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L12557-0P
L12557-22
L12557-36
CAAAAL12557-0PEA
251.22
CAD
CAAAAL12557-0P
CAAAAL12557-22
CAAAAL12557-36
Sodium dihydrogen citrate 99%
Sodium dihydrogen citrate
Purity: 99%
CAS No.: 18996-35-5
Molecular Formula: C6H7NaO7
Formula Weight: 214.11
Melting Point: 211-212°
Beilstein Reference: 3920246
Formula:
C₆H₇NaO₇ Melting Pt: 211…212 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013067 CAS Number: 18996-35-5 EINECS: 242-734-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...