(3-Phenoxyphenyl)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Phenoxyphenylacetic acid

L12524-03 L12524-06
CAAAAL12524-03EA 56.09 CAD
CAAAAL12524-03 CAAAAL12524-06
(3-Phenoxyphenyl)acetic acid 98%
(3-Phenoxyphenyl)acetic acid

Purity: 98% CAS No.: 32852-81-6 Molecular Formula: C14H12O3 Formula Weight: 228.25 Melting Point: 88-90° Beilstein Reference: 2730540

Formula: (C₆H₅OC₆H₅)CH₂COOH
Melting Pt: 88…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00016826
CAS Number: 32852-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR