2-Bromophenyl isocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L12521-03
L12521-06
L12521-14
CAAAAL12521-03EA
54.83
CAD
CAAAAL12521-03
CAAAAL12521-06
CAAAAL12521-14
2-Bromophenyl isocyanate 97%
2-Bromophenyl isocyanate
Purity: 97%
CAS No.: 1592-00-3
Molecular Formula: C7H4BrNO
Formula Weight: 198.03
Storage Temperature: 2°C to 8°C
Flash Point: 107°(224°F)
Refractive Index: 1.5850
Odor: Pungent
Boiling Point: 91-92°/5mm
Beilstein Reference: 3588447
UN No.: 2206
MDL No.: MFCD00001995
Formula:
C₇H₄BrNO Boiling Pt: 91…92 °C/5 mm Density: 1.607 Flash Pt: 107 °C (224 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00001995 CAS Number: 1592-00-3 UN: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...