1,1'-[Oxybis(ethyleneoxy)]diethylene 98% stabilised

Supplier: Thermo Scientific Chemicals

Synonyms: Diethylene glycol divinyl ether, 1,1'-[Oxybis(ethyleneoxy)]diethene, 1,1'-(Oxybis(ethyleneoxy))diethylene, 1,1'-(Oxybis(ethyleneoxy))diethene, Diethyleneglycoldivinylether, (2-(2-Ethenyloxyethoxy)ethoxy)ethene

Stabilization: 0.1% Stabilised with potassium hydroxide

L12501-09 L12501-18
CAAAAL12501-09EA 86.9 CAD
CAAAAL12501-09 CAAAAL12501-18
1,1'-[Oxybis(ethyleneoxy)]diethylene 98% stabilised
1,1'-[Oxybis(ethyleneoxy)]diethylene

Purity: 98% CAS No.: 764-99-8 Molecular Formula: C8H14O3 Formula Weight: 158.20 Flash Point: 83°(181°F) Refractive Index: 1.4470 Melting Point: -21° Boiling Point: 198-199° Beilstein Reference: 1752316 MDL No.: MFCD00054610

Formula: H₂CCHO(CH₂)₂O(CH₂)₂OCHCH₂
MW: 158.2 g/mol
Boiling Pt: 198…199 °C
Melting Pt: –21 °C
Density: 0.968
Flash Pt: 83 °C (181 °F)
Storage Temperature: Ambient
MDL Number: MFCD00054610
CAS Number: 764-99-8
EINECS: 212-133-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR