1-Bromo-3-phenoxypropane 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
3-Bromopropyl phenyl ether, (3-Bromopropoxy)benzene, 3-Phenoxypropyl bromide
L12494-14
L12494-22
CAAAAL12494-14EA
107.12
CAD
CAAAAL12494-14
CAAAAL12494-22
1-Bromo-3-phenoxypropane 98%
1-Bromo-3-phenoxypropane
Purity: 98%
CAS No.: 588-63-6
Molecular Formula: C6H5OCH2CH2CH2Br
Formula Weight: 215.10
Flash Point: 96°(204°F)
Refractive Index: 1.5460
Melting Point: 10-11°
Boiling Point: 130-134°/14mm
Beilstein Reference: 508978
MDL No.: MFCD00000256
Formula:
(C₆H₅)OCH₂CH₂CH₂Br MW: 215.09 g/mol Boiling Pt: 130…134 °C/14 mm Melting Pt: 10…11 °C Density: 1.365 Flash Pt: 96 °C(204 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000256 CAS Number: 588-63-6 EINECS: 209-623-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...