Tetradecyltrimethylammonium chloride 98%

Supplier: Thermo Scientific Chemicals
Warning

L12484-06 L12484-14
CAAAAL12484-06EA 87.69 CAD
CAAAAL12484-06 CAAAAL12484-14
Tetradecyltrimethylammonium chloride 98%
Tetradecyltrimethylammonium chloride

Purity: 98% CAS No.: 4574-04-3 Molecular Formula: CH3(CH2)13N(CH3)3Cl Formula Weight: 291.95 Beilstein Reference: 3916572 MDL No.: MFCD00059968

Formula: C₁₇H₃₈ClN
Storage Temperature: Ambient
MDL Number: MFCD00059968
CAS Number: 4574-04-3
EINECS: 224-958-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR