3-Bromobenzyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Bromo-α-chlorotoluene, 1-(Chloromethyl)-3-bromobenzene, α-Chloro-3-bromotoluene, 1-Bromo-3-(chloromethyl)benzene

L12458-06 L12458-14
CAAAAL12458-06EA 80.26 CAD
CAAAAL12458-06 CAAAAL12458-14
3-Bromobenzyl chloride 97%
3-Bromobenzyl chloride

Purity: 97% CAS No.: 932-77-4 Molecular Formula: C7H6BrCl Formula Weight: 205.49 Flash Point: 110°(230°F) Refractive Index: 1.5835 Boiling Point: 118-119°/18mm Beilstein Reference: 742267 UN No.: 3265 MDL No.: MFCD00040865

Formula: C₇H₆BrCl
Boiling Pt: 118…119 °C/18 mm
Melting Pt: 32.5 °C
Density: 1.608
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040865
CAS Number: 932-77-4
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR