L-Homoserine 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-HomoSer-OH, H-HoSer-OH, H-Hser-OH, (S)-(-)-Homoserine, L(-)-Homoserine
L12427-03
L12427-06
L12427-MD
CAAAAL12427-03EA
167.48
CAD
CAAAAL12427-03
CAAAAL12427-06
CAAAAL12427-MD
L-Homoserine 99%
L-Homoserine
Purity: 99%
CAS No.: 672-15-1
Molecular Formula: C4H9NO3
Formula Weight: 119.12
Melting Point: ca 203° dec.
Beilstein Reference: 1721681
Merck Reference: 14,4739
Optical Rotation: -9° (c=2 in water)
MDL No.: MFCD00063090
Formula:
C₄H₉NO₃ MW: 119.12 g/mol Melting Pt: 203 (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00063090 CAS Number: 672-15-1 EINECS: 211-590-6 Merck Index: 14,4739 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...