4'-Methyl-2,2,2-trifluoroacetophenone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,2,2-Trifluoro-1-(p-tolyl)ethanone, 4-(Trifluoroacetyl)toluene, 2,2,2-Trifluoro-1-(4-methylphenyl)ethan-1-one
L12424-03
L12424-06
CAAAAL12424-03EA
119.61
CAD
CAAAAL12424-03
CAAAAL12424-06
4'-Methyl-2,2,2-trifluoroacetophenone 97%
4'-Methyl-2,2,2-trifluoroacetophenone
Purity: 97%
CAS No.: 394-59-2
Molecular Formula: C9H7F3O
Formula Weight: 188.15
Refractive Index: 1.4700
Boiling Point: 70-72°/15mm
MDL No.: MFCD00052338
Formula:
C₉H₇F₃O MW: 188.15 Boiling Pt: 70…72 °C (15 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00052338 CAS Number: 394-59-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...