2'-Fluoropropiophenone 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L12423-04
L12423-09
L12423-18
CAAAAL12423-04EA
46.14
CAD
CAAAAL12423-04
CAAAAL12423-09
CAAAAL12423-18
2'-Fluoropropiophenone 99%
2'-Fluoropropiophenone
Purity: 99%
CAS No.: 446-22-0
Molecular Formula: C9H9FO
Formula Weight: 152.17
Flash Point: 79°(174°F)
Refractive Index: 1.5045
Boiling Point: 89-91°/12mm
Beilstein Reference: 3235723
MDL No.: MFCD00009893
Formula:
C₉H₉FO Boiling Pt: 89…91 °C/12 mm Density: 1,102 Flash Pt: 79 °C(174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009893 CAS Number: 446-22-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...