3,5-Di-tert-butylbenzoic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L12386-03
L12386-06
CAAAAL12386-03EA
33.97
CAD
CAAAAL12386-03
CAAAAL12386-06
3,5-Di-tert-butylbenzoic acid 99%
3,5-Di-tert-butylbenzoic acid
Purity: 99%
CAS No.: 16225-26-6
Molecular Formula: C15H22O2
Formula Weight: 234.34
Melting Point: 160-162°
Beilstein Reference: 981248
Formula:
C₁₅H₂₂O₂ Melting Pt: 160…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082727 CAS Number: 16225-26-6 EINECS: 240-350-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...