2-Chloropropene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
α-Methylvinyl chloride
L12374-06
L12374-14
CAAAAL12374-06EA
116.45
CAD
CAAAAL12374-06
CAAAAL12374-14
2-Chloropropene 99%
2-Chloropropene
Purity: 99%
CAS No.: 557-98-2
Molecular Formula: CH3CCl=CH2
Formula Weight: 76.53
Storage Temperature: 2°C to 8°C
Flash Point: -34°(-29°F)
Refractive Index: 1.3939
Odor: Recognizable
Melting Point: -138°
Boiling Point: 22-23°
Beilstein Reference: 1361376
UN No.: 2456
MDL No.: MFCD00000859
Formula:
C₃H₅Cl MW: 76.53 g/mol Boiling Pt: 22…23 °C Melting Pt: –138 °C Density: 0.899 Flash Pt: –34 °C (–29 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000859 CAS Number: 557-98-2 EINECS: 209-187-5 UN: 2456 ADR: 3,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...