4-Methoxybenzyl isothiocyanate 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L12342-03
L12342-06
CAAAAL12342-03EA
72.84
CAD
CAAAAL12342-03
CAAAAL12342-06
4-Methoxybenzyl isothiocyanate 94%
4-Methoxybenzyl isothiocyanate
Purity: 94%
CAS No.: 3694-57-3
Molecular Formula: C9H9NOS
Formula Weight: 179.24
Refractive Index: 1.5975
Odor: Irritating
Boiling Point: 132-133°/0.7mm
Beilstein Reference: 2089549
UN No.: 2922
MDL No.: MFCD00041117
Formula:
C₉H₉NOS Boiling Pt: 132…133 °C/0.7 mm Density: 1.150 Storage Temperature: Ambient |
MDL Number:
MFCD00041117 CAS Number: 3694-57-3 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...