tert-Butanol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L12338-0F
L12338-AK
L12338-AU
CAAAAL12338-0FEA
159.58
CAD
CAAAAL12338-0F
CAAAAL12338-AK
CAAAAL12338-AU
tert-Butanol 99%
tert-Butanol
Purity: 99%
CAS No.: 75-65-0
Molecular Formula: (CH3)3COH
Formula Weight: 74.12
Flash Point: 11°(52°F)
Refractive Index: 1.3870
Melting Point: 25°
Boiling Point: 83°
Beilstein Reference: 906698
Merck Reference: 14,1542
UN No.: 1120
MDL No.: MFCD00004464
Formula:
(CH₃)₃COH Boiling Pt: 83 °C Melting Pt: 25 °C Density: 0.786 Flash Pt: 11 °C (52 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004464 CAS Number: 75-65-0 EINECS: 200-889-7 UN: 1120 ADR: 3,III Merck Index: 14,1542 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...