o-Tolyl isocyanate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L12247-06
L12247-14
CAAAAL12247-06EA
68.89
CAD
CAAAAL12247-06
CAAAAL12247-14
o-Tolyl isocyanate 98%
o-Tolyl isocyanate
Purity: 98%
CAS No.: 614-68-6
Molecular Formula: C8H7NO
Formula Weight: 133.15
Storage Temperature: 2°C to 8°C
Flash Point: 92°(197°F)
Refractive Index: 1.5345
Odor: Pungent
Boiling Point: 186°
Beilstein Reference: 507985
UN No.: 2206
MDL No.: MFCD00002009
Formula:
C₈H₇NO Boiling Pt: 186 °C Density: 1.074 Flash Pt: 92 °C (197 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002009 CAS Number: 614-68-6 EINECS: 210-389-0 UN: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...