4,5-Dicyanoimidazole 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4,5-Dicyano-1H-imidazole, Imidazole-4,5-dicarbonitrile

L12215-06 L12215-14
CAAAAL12215-06EA 57.99 CAD
CAAAAL12215-06 CAAAAL12215-14
4,5-Dicyanoimidazole 98+%
4,5-Dicyanoimidazole

Purity: 98+% CAS No.: 1122-28-7 Molecular Formula: C5H2N4 Formula Weight: 118.10 Odor: Odorless Melting Point: 173-177° Beilstein Reference: 118208 MDL No.: MFCD00005194

Formula: C₅H₂N₄
MW: 118.1 g/mol
Melting Pt: 176
Storage Temperature: Ambient
MDL Number: MFCD00005194
CAS Number: 1122-28-7
EINECS: 214-344-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR