4-Nitro-m-phenylenediamine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L12166-09
L12166-18
CAAAAL12166-09EA
58.3
CAD
CAAAAL12166-09
CAAAAL12166-18
4-Nitro-m-phenylenediamine 95%
4-Nitro-m-phenylenediamine
Purity: 95%
CAS No.: 5131-58-8
Molecular Formula: C6H7N3O2
Formula Weight: 153.14
Melting Point: 156-160°
Beilstein Reference: 2094904
Formula:
C₆H₇N₃O₂ Melting Pt: 156…160 °C Density: 1.552 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00025289 CAS Number: 5131-58-8 EINECS: 225-876-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...