2-Methoxy-4-nitrophenyl isothiocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L12150-03
L12150-06
CAAAAL12150-03EA
46.93
CAD
CAAAAL12150-03
CAAAAL12150-06
2-Methoxy-4-nitrophenyl isothiocyanate 97%
2-Methoxy-4-nitrophenyl isothiocyanate
Purity: 97%
CAS No.: 190774-55-1
Molecular Formula: C8H6N2O3S
Formula Weight: 210.21
Melting Point: 107-109°
Beilstein Reference: 6924925
UN No.: 2923
MDL No.: MFCD00041379
Formula:
C₈H₆N₂O₃S MW: 210.21 g/mol Melting Pt: 107…109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041379 CAS Number: 190774-55-1 UN: 2923 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...