Dimethyldichlorosilane 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dichloro(dimethyl)silane, Dichlorodimethylsilane

L12133-0B L12133-22 L12133-36
CAAAAL12133-0BEA 161.16 CAD
CAAAAL12133-0B CAAAAL12133-22 CAAAAL12133-36
Dimethyldichlorosilane 99%
Dimethyldichlorosilane

Purity: 99% CAS No.: 75-78-5 Molecular Formula: C2H6Cl2Si Formula Weight: 129.06 Flash Point: -5°(23°F) Refractive Index: 1.4045 Melting Point: ca -76° Boiling Point: 70-71° Beilstein Reference: 605287 UN No.: 1162 MDL No.: MFCD00000491

Formula: C₂H₆Cl₂Si
MW: 129.06 g/mol
Boiling Pt: 70…71 °C
Melting Pt: ∼ 76 °C
Density: 1.068
Flash Pt: –5 °C (23 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000491
CAS Number: 75-78-5
EINECS: 200-901-0
UN: 1162
ADR: 3,II
REACH: 01-2119437250-51

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR