5-Chloro-1-pentanol 95%

Supplier: Thermo Scientific Chemicals
Warning

L12088-09 L12088-18
CAAAAL12088-09EA 147.26 CAD
CAAAAL12088-09 CAAAAL12088-18
5-Chloro-1-pentanol 95%
5-Chloro-1-pentanol

Purity: 95% CAS No.: 5259-98-3 Molecular Formula: Cl(CH2)5OH Formula Weight: 122.60 Refractive Index: 1.4540 Boiling Point: 102-103°/8mm MDL No.: MFCD00039556

Formula: C₅H₁₁ClO
Boiling Pt: 102…103 °C (8 mmHg)
Density: 1.060
Storage Temperature: Ambient
MDL Number: MFCD00039556
CAS Number: 5259-98-3
EINECS: 226-067-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR