1-Bromobutanone tech. 90% stabilized, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Bromo-2-butanone
Stabilization:
Stabilised with calcium carbonate
L12081-03
L12081-06
CAAAAL12081-03EA
104.6
CAD
CAAAAL12081-03
CAAAAL12081-06
1-Bromobutanone tech. 90% stabilized, Technical Grade
1-Bromobutanone
Purity: 90%
CAS No.: 816-40-0
Molecular Formula: C4H7BrO
Formula Weight: 151.01
Flash Point: 68°(154°F)
Refractive Index: 1.4650
Odor: Pungent
Boiling Point: 48-49°/10mm
Beilstein Reference: 741894
UN No.: 2927
MDL No.: MFCD00000207
Formula:
BrH₂CC(O)CH₂CH₃ MW: 151 g/mol Boiling Pt: 48…49 °C/10 mm Density: 1.479 Flash Pt: 68 °C(154 °F) Storage Temperature: Freezer |
MDL Number:
MFCD00000207 CAS Number: 816-40-0 EINECS: 212-431-3 UN: 3334 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...