3-Chloro-4-methylphenyl isocyanate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L12064-03
L12064-06
CAAAAL12064-03EA
35.39
CAD
CAAAAL12064-03
CAAAAL12064-06
3-Chloro-4-methylphenyl isocyanate 98%
3-Chloro-4-methylphenyl isocyanate
Purity: 98%
CAS No.: 28479-22-3
Molecular Formula: C8H6ClNO
Formula Weight: 167.60
Storage Temperature: 2°C to 8°C
Flash Point: 109°(228°F)
Refractive Index: 1.5560
Melting Point: 20-24°
Boiling Point: 225°
Beilstein Reference: 2690771
UN No.: 2236
MDL No.: MFCD00013858
Formula:
C₈H₆ClNO Boiling Pt: 225 °C Melting Pt: 20…24 °C Density: 1.224 Flash Pt: 109 °C (228 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00013858 CAS Number: 28479-22-3 EINECS: 249-050-7 UN: 3428 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...