3,4-Dichlorothiophenol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2-Dichloro-4-mercaptobenzene, 1,2-Dichloro-4-benzenethiol, 3,4-Dichlorobenzenethiol, 3,4-Dichlorobenzene-1-thiol

L12031-03 L12031-06
CAAAAL12031-03EA 39.5 CAD
CAAAAL12031-03 CAAAAL12031-06
3,4-Dichlorothiophenol 97%
3,4-Dichlorothiophenol

Purity: 97% CAS No.: 5858-17-3 Molecular Formula: C6H4Cl2S Formula Weight: 179.07 Flash Point: 110°(230°F) Refractive Index: 1.6230 Odor: Unpleasant Boiling Point: 119-120°/10mm Beilstein Reference: 2042869 MDL No.: MFCD00004840

Formula: C₆H₄Cl₂S
MW: 179.07 g/mol
Boiling Pt: 119…120 °C (10 mmHg)
Melting Pt: 87…89 °C
Density: 1.407
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004840
CAS Number: 5858-17-3
EINECS: 227-485-3
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR