3-(Trifluoromethyl)phenylhydrazine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L11996-03
L11996-06
CAAAAL11996-03EA
79.63
CAD
CAAAAL11996-03
CAAAAL11996-06
3-(Trifluoromethyl)phenylhydrazine 95%
3-(Trifluoromethyl)phenylhydrazine
Purity: 95%
CAS No.: 368-78-5
Molecular Formula: C7H7F3N2
Formula Weight: 176.14
Flash Point: 107°(224°F)
Refractive Index: 1.5040
Boiling Point: 80-83°/9mm
Beilstein Reference: 640075
MDL No.: MFCD00025093
Formula:
C₇H₇F₃N₂ Boiling Pt: 80…83 °C (9 mmHg) Density: 1.348 Flash Pt: 107 °C (224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00025093 CAS Number: 368-78-5 EINECS: 206-713-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...