EDTA tripotassium salt dihydrate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethylenedinitrilotetra-acetic acid tripotassium salt dihydrate, Tripotassium hydrogen ethylenediaminetetraacetate dihydrate, Ethylenediaminetetra-acetic acid tripotassium salt dihydrate, Potassium 2,2'-((2-((carboxylatomethyl)(carboxymethyl)amino)ethyl)azanediyl)diacetate dihydrate, Edetic acid tripotassium salt dihydrate
Purity: 99%
CAS No.: 65501-24-8
Molecular Formula: C10H13K3N2O8.2H2O
Formula Weight: 442.55 (406.46anhy)
Melting Point: ca 182° dec.
Beilstein Reference: 6762497
Formula:
C₁₀H₁₃K₃N₂O₈·2H₂O MW: 442.55 g/mol Melting Pt: ∼ 182 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00149672 CAS Number: 65501-24-8 EINECS: 241-543-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...