Cyclopropyl isothiocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L11985-03
L11985-06
CAAAAL11985-03EA
87.37
CAD
CAAAAL11985-03
CAAAAL11985-06
Cyclopropyl isothiocyanate 97%
Cyclopropyl isothiocyanate
Purity: 97%
CAS No.: 56601-42-4
Molecular Formula: C4H5NS
Formula Weight: 99.16
Storage Temperature: 2°C to 8°C
Flash Point: 57°(134°F)
Refractive Index: 1.5480
Boiling Point: 87-88°/35mm
Beilstein Reference: 3600974
UN No.: 2929
MDL No.: MFCD00040874
Formula:
C₄H₅NS Boiling Pt: 87…88 °C/35 mm Density: 1.070 Flash Pt: 57 °C (134 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00040874 CAS Number: 56601-42-4 EINECS: 260-278-6 UN: 3286 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...