2-Bromobenzyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L11953-03
L11953-06
CAAAAL11953-03EA
56.41
CAD
CAAAAL11953-03
CAAAAL11953-06
2-Bromobenzyl chloride 97%
2-Bromobenzyl chloride
Purity: 97%
CAS No.: 578-51-8
Molecular Formula: C7H6BrCl
Formula Weight: 205.49
Refractive Index: 1.5860
Boiling Point: 110-111°/15mm
UN No.: 3265
MDL No.: MFCD00040863
Formula:
C₇H₆BrCl Boiling Pt: 110…111 °C/15 mm Density: 1.7341 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00040863 CAS Number: 578-51-8 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...