3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Thiazolyl blue tetrazolium bromide, MTT, 2-(4,5-Dimethylthiazol-2-yl)-3,5-diphenyl-2H-tetrazol-3-ium bromide

L11939-03 L11939-06
CAAAAL11939-03EA 111.07 CAD
CAAAAL11939-03 CAAAAL11939-06
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) 98%
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)

Used in measurement of cell proliferation. Produces a yellowish solution that is converted to dark blue, water-insoluble MTT formazan by mitochondrial dehydrogenases of living cells


Purity: 98% CAS No.: 298-93-1 Molecular Formula: C18H16BrN5S Formula Weight: 414.33 Odor: Odorless Melting Point: ca 195° dec. Beilstein Reference: 3825277 MDL No.: MFCD00011964

Formula: C₁₈H₁₆N₅BrS
MW: 414.33 g/mol
Melting Pt: ∼ 195 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011964
CAS Number: 298-93-1
EINECS: 206-069-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR