4-(Trifluoromethyl)phenyl isothiocyanate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L11890-03
L11890-06
CAAAAL11890-03EA
63.99
CAD
CAAAAL11890-03
CAAAAL11890-06
4-(Trifluoromethyl)phenyl isothiocyanate 98+%
4-(Trifluoromethyl)phenyl isothiocyanate
Purity: 98+%
CAS No.: 1645-65-4
Molecular Formula: C8H4F3NS
Formula Weight: 203.19
Flash Point:
Formula:
C₈H₄F₃NS Boiling Pt: 207 °C Melting Pt: 39…42 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039645 CAS Number: 1645-65-4 UN: 2923 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...