5-Nitro-o-toluic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methyl-5-nitrobenzoic acid

L11862-06 L11862-14
CAAAAL11862-03EA 77.56 CAD
CAAAAL11862-03 CAAAAL11862-06 CAAAAL11862-14
5-Nitro-o-toluic acid 98+%
5-Nitro-o-toluic acid

Purity: 98+% CAS No.: 1975-52-6 Molecular Formula: C8H7NO4 Formula Weight: 181.15 Melting Point: 176-181° Beilstein Reference: 2451300

Formula: C₈H₇NO₄
MW: 181.15
Melting Pt: 176…181 °C
Density: 1.49
Storage Temperature: Ambient
MDL Number: MFCD00007371
CAS Number: 1975-52-6
EINECS: 217-829-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR