Charcoal activated, powder, Norit® GSX, steam activated and acid washed

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Carbon activated, Activated carbon, Activated charcoal

L11860-0P L11860-22 L11860-36
CAAAAL11860-0PEA 540.36 CAD
CAAAAL11860-0P CAAAAL11860-22 CAAAAL11860-36
Charcoal activated, powder, Norit® GSX, steam activated and acid washed
Charcoal activated

Decolorizing carbon


CAS No.: 7440-44-0 UN No.: 1362 MDL No.: MFCD00133992

Formula: C
Melting Pt: 3550 °C
Density: 2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1362
ADR: 4.2,III
REACH: 01-2119488894-16
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR