Ethylenediaminetetraacetic acid disodium magnesium salt hydrate

Supplier: Thermo Scientific Chemicals
Danger

L11857-14 L11857-22
CAAAAL11857-14EA 50.09 CAD
CAAAAL11857-14 CAAAAL11857-22
Ethylenediaminetetraacetic acid disodium magnesium salt hydrate
Ethylenediaminetetraacetic acid disodium magnesium salt hydrate

Ethylenediaminetetraacetic acid disodium magnesium salt hydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is used as chelating agents over a broad pH range in aqueous systems.


CAS No.: 14402-88-1 Molecular Formula: C10H12MgN2Na2O8.xH2O Formula Weight: 358.50(anhy) Odor: Odorless Melting Point: 300° MDL No.: MFCD00078217

Formula: C₁₀H₁₂MgN₂Na₂O₈·xH₂O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00078217
CAS Number: 194491-32-2
EINECS: 238-372-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR