Dibenzyl disulfide 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzyl disulphide, Benzyl disulfide

L11850-14 L11850-22
CAAAAL11850-14EA 42.66 CAD
CAAAAL11850-14 CAAAAL11850-22
Dibenzyl disulfide 98+%
Dibenzyl disulphide

Purity: 98+% CAS No.: 150-60-7 Molecular Formula: C6H5CH2S-SCH2C6H5 Formula Weight: 246.40 Flash Point: 150°(302°F) Odor: Stench Boiling Point: 210-216°/18mm Beilstein Reference: 1110443 Merck Reference: 14,3013 MDL No.: MFCD00004783

Formula: C₁₄H₁₄S₂
MW: 246.4 g/mol
Boiling Pt: 210…216 °C/18 mm
Melting Pt: 67…72 °C
Density: 1.300
Flash Pt: 150 °C(302 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004783
CAS Number: 150-60-7
EINECS: 205-764-0
Merck Index: 14,3013

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR