10-Undecin-1-ol 96%

Supplier: Thermo Scientific Chemicals

Synonyms: 10-Undecyn-1-ol, Undec-10-yn-1-ol, 1-Undecyn-11-ol

L11807-03 L11807-06
CAAAAL11807-03EA 92.59 CAD
CAAAAL11807-03 CAAAAL11807-06
10-Undecin-1-ol 96%
10-Undecin-1-ol

Purity: 96% CAS No.: 2774-84-7 Molecular Formula: HC≡C(CH2)9OH Formula Weight: 168.28 Refractive Index: 1.4565 Boiling Point: 110-111°/4mm MDL No.: MFCD00041675

Formula: C₁₁H₂₀O
MW: 168.28 g/mol
Boiling Pt: 110…111 °C (4 mmHg)
Density: 0.873
Storage Temperature: Ambient
MDL Number: MFCD00041675
CAS Number: 2774-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR