Dithiodiglycol (bis(2-hydroxyethyl) disulfide) tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Danger

L11800-18 L11800-30
CAAAAL11800-18EA 96.54 CAD
CAAAAL11800-18 CAAAAL11800-30
Dithiodiglycol (bis(2-hydroxyethyl) disulfide) tech. 90%, Technical Grade
Dithiodiglycol (Bis(2-hydroxyethyl) disulphide)

Purity: 90% CAS No.: 1892-29-1 Molecular Formula: C4H10O2S2 Formula Weight: 154.24 Flash Point: 110°(230°F) Refractive Index: 1.5700 Odor: Unpleasant Melting Point: ca 25° Boiling Point: 158-163°/3.5mm Beilstein Reference: 1735851 UN No.: 2810 MDL No.: MFCD00002906

Formula: C₄H₁₀O₂S₂
Boiling Pt: 158…163 °C/3.5 mm
Melting Pt: ∼ 25 °C
Density: 1.261
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002906
CAS Number: 1892-29-1
EINECS: 217-576-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR