2-Methoxyphenyl isothiocyanate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L11799-06
L11799-14
CAAAAL11799-06EA
92.75
CAD
CAAAAL11799-06
CAAAAL11799-14
2-Methoxyphenyl isothiocyanate 98%
2-Methoxyphenyl isothiocyanate
Purity: 98%
CAS No.: 3288-04-8
Molecular Formula: C8H7NOS
Formula Weight: 165.22
Flash Point:
Formula:
C₈H₇NOS Boiling Pt: 267 °C Density: 1.196 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011675 CAS Number: 3288-04-8 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...