trans-5-Decen-1-ol 96%
Supplier: Thermo Scientific Chemicals
Warning
|
L11768-03
L11768-06
CAAAAL11768-03EA
93.38
CAD
CAAAAL11768-03
CAAAAL11768-06
trans-5-Decen-1-ol 96%
trans-5-Decen-1-ol
Purity: 96%
CAS No.: 56578-18-8
Molecular Formula: C10H20O
Formula Weight: 156.27
Flash Point: 62°(143°F)
Refractive Index: 1.4520
Boiling Point: 113-118°/11mm
Beilstein Reference: 1840630
MDL No.: MFCD00056187
Formula:
C₁₀H₂₀O Boiling Pt: 113…118 °C (11 mmHg) Flash Pt: 62 °C (143 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00056187 CAS Number: 56578-18-8 EINECS: 260-267-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...