1-Chloro-3,3-dimethylbutane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L11753-09
L11753-18
CAAAAL11753-09EA
117.39
CAD
CAAAAL11753-09
CAAAAL11753-18
1-Chloro-3,3-dimethylbutane 97%
1-Chloro-3,3-dimethylbutane
Purity: 97%
CAS No.: 2855-08-5
Molecular Formula: C6H13Cl
Formula Weight: 120.62
Flash Point: 7°(44°F)
Refractive Index: 1.4170
Boiling Point: 116-118°
Beilstein Reference: 1731465
UN No.: 1993
MDL No.: MFCD00018980
Formula:
C₆H₁₃Cl Boiling Pt: 116…118 °C Density: 0.867 Flash Pt: 7 °C (44 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00018980 CAS Number: 2855-08-5 EINECS: 220-665-2 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...