(4-Fluorophenyl)acetone 99%
Supplier: Thermo Scientific Chemicals
Warning
|
L11720-03
L11720-06
CAAAAL11720-03EA
54.98
CAD
CAAAAL11720-03
CAAAAL11720-06
(4-Fluorophenyl)acetone 99%
(4-Fluorophenyl)acetone
Purity: 99%
CAS No.: 459-03-0
Molecular Formula: C9H9FO
Formula Weight: 152.17
Flash Point: 91°(195°F)
Refractive Index: 1.4958
Boiling Point: 106-107°/18mm
Beilstein Reference: 1936697
MDL No.: MFCD00000362
Formula:
C₉H₉FO Boiling Pt: 106…107 °C/18 mm Density: 1.11 Flash Pt: 91 °C(195 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000362 CAS Number: 459-03-0 EINECS: 207-284-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...