3,5-Dimethylphenyl isocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3,5-Xylyl isocyanate, 3,5-Dimethylphenylisocyanate
L11698-03
L11698-06
CAAAAL11698-03EA
42.19
CAD
CAAAAL11698-03
CAAAAL11698-06
3,5-Dimethylphenyl isocyanate 97%
3,5-Dimethylphenyl isocyanate
Purity: 97%
CAS No.: 54132-75-1
Molecular Formula: C9H9NO
Formula Weight: 147.18
Storage Temperature: 2°C to 8°C
Flash Point: 83°(181°F)
Refractive Index: 1.5280
Boiling Point: 205°
Beilstein Reference: 774958
UN No.: 2206
MDL No.: MFCD00013868
Formula:
C₉H₉NO Boiling Pt: 205 °C Density: 1.045 Flash Pt: 83 °C (181 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00013868 CAS Number: 54132-75-1 EINECS: 258-987-0 UN: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...