5-Chloro-7-methylisatin 97%
Supplier: Thermo Scientific Chemicals
L11682-03
L11682-06
L11682-MD
CAAAAL11682-03EA
456.62
CAD
CAAAAL11682-03
CAAAAL11682-06
CAAAAL11682-MD
5-Chloro-7-methylisatin 97%
5-Chloro-7-methylisatin
Purity: 97%
CAS No.: 14389-06-1
Molecular Formula: C9H6ClNO2
Formula Weight: 195.61
Melting Point: ca 283° dec.
Beilstein Reference: 153061
Formula:
C₉H₆ClNO₂ Melting Pt: ∼ 283 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00047218 CAS Number: 14389-06-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...