3-(Trifluoromethyl)phenyl isothiocyanate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L11646-03
L11646-06
L11646-14
CAAAAL11646-03EA
39.18
CAD
CAAAAL11646-03
CAAAAL11646-06
CAAAAL11646-14
3-(Trifluoromethyl)phenyl isothiocyanate 98%
3-(Trifluoromethyl)phenyl isothiocyanate
Purity: 98%
CAS No.: 1840-19-3
Molecular Formula: C8H4F3NS
Formula Weight: 203.19
Flash Point: 95°(203°F)
Refractive Index: 1.5547
Odor: Pungent
Boiling Point: 208°
Beilstein Reference: 781205
UN No.: 2922
MDL No.: MFCD00004806
Formula:
C₈H₄F₃NS Boiling Pt: 208 °C Density: 1.335 Flash Pt: 95 °C (203 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004806 CAS Number: 1840-19-3 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...