2-Chloro-6-methylphenyl isocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L11645-03
L11645-06
CAAAAL11645-03EA
42.98
CAD
CAAAAL11645-03
CAAAAL11645-06
2-Chloro-6-methylphenyl isocyanate 97%
2-Chloro-6-methylphenyl isocyanate
Purity: 97%
CAS No.: 40398-01-4
Molecular Formula: C8H6ClNO
Formula Weight: 167.60
Storage Temperature: 2°C to 8°C
Flash Point: 107°(224°F)
Refractive Index: 1.5570
Boiling Point: 79-80°/2mm
Beilstein Reference: 2690768
UN No.: 2206
MDL No.: MFCD00037030
Formula:
C₈H₆ClNO Boiling Pt: 79…80 °C/2 mm Density: 1.231 Flash Pt: 107 °C (224 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00037030 CAS Number: 40398-01-4 UN: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...