N-Phenyl-o-phenylenediamine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(2-Aminophenyl)aniline, 2-Aminodiphenylamine, N1-Phenylbenzene-1,2-diamine

L11616-06 L11616-14
CAAAAL11616-06EA 56.09 CAD
CAAAAL11616-06 CAAAAL11616-14
N-Phenyl-o-phenylenediamine 98%
N-Phenyl-o-phenylenediamine

Purity: 98% CAS No.: 534-85-0 Molecular Formula: C12H12N2 Formula Weight: 184.24 Melting Point: 77-80° Beilstein Reference: 908984

Formula: C₁₂H₁₂N₂
Boiling Pt: 106…108 °C (0.4 mmHg)
Melting Pt: 77…80 °C
Density: 1.234 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007685
CAS Number: 534-85-0
EINECS: 208-606-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR