4-Pentynylbenzene 98+%
Supplier: Thermo Scientific Chemicals
Warning
|
L11608-06
L11608-14
CAAAAL11608-06EA
134.46
CAD
CAAAAL11608-06
CAAAAL11608-14
4-Pentynylbenzene 98+%
4-Pentynylbenzene
Purity: 98+%
CAS No.: 1823-14-9
Molecular Formula: C11H12
Formula Weight: 144.22
Flash Point: 85°(185°F)
Refractive Index: 1.5165
Boiling Point: 94°/50mm
Beilstein Reference: 2074729
MDL No.: MFCD00039813
Formula:
(C₆H₅)CH₂CH₂CH₂CCH Boiling Pt: 94 °C/50 mm Density: 0.950 Flash Pt: 85 °C(185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039813 CAS Number: 1823-14-9 EINECS: 217-352-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...