2-Chloroethyl isothiocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L11604-03
L11604-06
CAAAAL11604-03EA
40.76
CAD
CAAAAL11604-03
CAAAAL11604-06
2-Chloroethyl isothiocyanate 97%
2-Chloroethyl isothiocyanate
Purity: 97%
CAS No.: 6099-88-3
Molecular Formula: ClCH2CH2NCS
Formula Weight: 121.59
Flash Point: 96°(205°F)
Refractive Index: 1.5560
Boiling Point: 79-80°/13mm
Beilstein Reference: 605585
UN No.: 2922
MDL No.: MFCD00041125
Formula:
C₃H₄ClNS Boiling Pt: 79…80 °C/13 mm Density: 1.265 Flash Pt: 96 °C (205 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041125 CAS Number: 6099-88-3 UN: 2929 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...