1-tert-Butyl-4-iodobenzene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L11603-03
L11603-06
CAAAAL11603-03EA
78.37
CAD
CAAAAL11603-03
CAAAAL11603-06
1-tert-Butyl-4-iodobenzene 97%
1-tert-Butyl-4-iodobenzene
Purity: 97%
CAS No.: 35779-04-5
Molecular Formula: C10H13I
Formula Weight: 260.12
Flash Point: 106°(223°F)
Refractive Index: 1.5710
Boiling Point: 243°
Beilstein Reference: 1931810
MDL No.: MFCD00052339
Formula:
C₁₀H₁₃I Boiling Pt: 243 °C Density: 1,468 Flash Pt: 106 °C(223 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00052339 CAS Number: 35779-04-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...